Learn More
Mycoplasma bovis Mouse anti-Bacteria, Alexa Fluor™ 594, Clone: 10F181, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP230167AF594
Catalog No. NP230167A94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mycoplasma bovis Monoclonal antibody specifically detects Mycoplasma bovis in Bacteria samples. It is validated for ELISA.
Specifications
Specifications
Mycoplasma bovis
10F181
ELISA
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Bacteria
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
M. bovis preparation
0.1 mL
Primary
Recognizes Mycoplasma bovis. Does not react with Streptococcus agalactaie, S. aureus, E. coli, M. bovigentalium or M. californcum.