Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycoplasma pneumoniae P1 Monoclonal Antibody (6536), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA547573

Catalog No. PIMA547573


Add to Cart

Description

Description

Specificity: Specific for the P1 adhesin of M. pneumoniae. Does not cross react with: Ureaplasma urealyticum, Mycoplasma hominis or Mycoplasma genitalium. Best ELISA pairs (capture/conjugate): Mycoplasma pneumoniae P1 Monoclonal antibody (Product # MA5-47571)/Mycoplasma pneumoniae P1 Monoclonal antibody (Product # MA5-47573). Purified preparations consist of >90% pure mouse MAb which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Mycoplasma pneumoniae P1 Monoclonal antibody specifically detects Mycoplasma pneumoniae P1 in Bacteria samples. It is validated for ELISA, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mycoplasma pneumoniae P1
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide, pH 7.2
Proprietary sequence
100 μg
Primary
4° C
IgG2a
ELISA, Immunocytochemistry
6536
Unconjugated
Mouse
Protein A
RUO
Bacteria
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit