Learn More
MYD88 Mouse anti-Human, Clone: OTI1B4, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA502104
Catalog No. 50-167-3187
Add to cart
Description
Description
This gene encodes a cytosolic adapter protein that plays a central role in the innate and adaptive immune response. This protein functions as an essential signal transducer in the interleukin-1 and Toll-like receptor signaling pathways. These pathways regulate that activation of numerous proinflammatory genes. The encoded protein consists of an N-terminal death domain and a C-terminal Toll-interleukin1 receptor domain. Patients with defects in this gene have an increased susceptibility to pyogenic bacterial infections. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
MYD88
Monoclonal
Unconjugated
MYD88
Liquid
MYD88
Full length human recombit protein of human MYD88 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1B4
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q99836
MYD88D
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4615