Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MyD88 Mouse anti-Human, DyLight™ 755, Clone: OTI2B2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272842IR
Description
MyD88 Monoclonal specifically detects MyD88 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MyD88 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MYD88 | |
Full length recombinant protein of human MyD88 (NP_002459) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2B2 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
Mouse | |
Affinity Purified | |
Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
4615.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title