Learn More
MyD88 Mouse anti-Human, Mouse, Primate, Rhesus Macaque, Alexa Fluor™ 350, Clone: 4D6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227369AF350
Catalog No. NP227369A30
Add to cart
Description
Description
MyD88 Monoclonal antibody specifically detects MyD88 in Human,Mouse,Primate,Rhesus Macaque samples. It is validated for Western Blot,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Flow Cytometry.
Specifications
Specifications
MyD88
4D6
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular)
MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
The immunogen is 100% homologous to isoform 3: 264 aa, isoform 4: 204 aa; isoform 5: 159 aa, CRA_a: 304 aa, CRA_b: 309 aa),
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
MYD88
A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody.
0.1 mL
Primary
4615
Human, Mouse, Primate, Rhesus Macaque