Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYEF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552128
Description
Immunogen sequence: GMGSMNSVTG GMGMGLDRMS SSFDRMGPGI GAILERSIDM DRGFLSGPMG SGMRERIGSK GNQIFVRNLP FDLTWQKLKE KFSQCGHVMF AEIKMENGKS KGCGTVRFDS PESAEKACRI MNGIKISGRE IDVRLDRN Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
MEF-2 is expressed early in the differentiation program and is suppressed by specific polypeptide growth factors. The ability of MEF-2 to recognize conserved activating elements associated with multiple-specific genes suggests that this factor may participate in the coordinate regulation of genes during myogenesis.
Specifications
MYEF2 | |
Polyclonal | |
Unconjugated | |
MYEF2 | |
9430071B01; HsT18564; hypothetical protein LOC641483; Kiaa1341; LOW QUALITY PROTEIN: myelin expression factor 2; Mef2; MEF-2; mKIAA1341; MST156; MSTP156; Myef2; myEF-2; myelin basic protein expression factor 2, repressor; myelin expression factor 2; myelin gene expression factor 2; zgc:123351; zgc:152722 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2K5 | |
MYEF2 | |
Recombinant protein corresponding to Human MYEF2. Recombinant protein control fragment (Product #RP-102488). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction