Learn More
Alfa Aesar™ Myelin Basic Protein (1-11), Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66051LB0
 View more versions of this product
Catalog No. AAJ66051LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1293.42
Human
Solid
RUO
Myelin Basic Protein (1-11), human
HPLC
5mg
N-Acetyl-Ala-Ser-Gln-Lys-Arg-Pro-Ser-Gln-Arg-His-Gly
-30° to -10°C