Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin PLP Mouse anti-Human, Mouse, Rat, Bovine, Reptile, Clone: PLP1/4259, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Myelin PLP
Clone PLP1/4259
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122679
View Documents
Novus Biologicals
NBP30760310UG
100 μg
Each for $352.89
Add to cart
 
NB122680
View Documents
Novus Biologicals
NBP30760320UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Myelin PLP Monoclonal antibody specifically detects Myelin PLP in Human, Mouse, Rat, Bovine, Reptile samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Myelin PLP
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse, Rat, Bovine, Reptile
HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2
Synthetic peptide GRGTKF that recognizes the C terminal region (aa272-277) of Myelin PLP.
Monoclonal
Protein A or G purified
PLP1/4259
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers
10 mM PBS with 0.05% BSA
5354
IgG2a
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit