Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloid-Associated Differentiation Marker, Rabbit anti-Human,Rat, PE, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224494PE

Catalog No. NB005257

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Myeloid-Associated Differentiation Marker Polyclonal antibody specifically detects Myeloid-Associated Differentiation Marker in Human, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

Myeloid-Associated Differentiation Marker
PE
PBS with 0.05% Sodium Azide
MYADM, Myeloid Upregulated Protein, Protein SB135, SB135
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Flow Cytometry
Flow Cytometry
Affinity Purified, Conjugated
MYADM
A portion of amino acids 250-300 of human MYADM was used as the immunogen for this antibody.
Immunogen affinity purified
RUO
Primary
Human, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit