Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloid Cell Marker Mouse anti-Human, FITC, Clone: BM-2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314086F
This item is not returnable.
View return policy
Description
Myeloid Cell Marker Monoclonal antibody specifically detects Myeloid Cell Marker in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Myeloid Cell Marker | |
Monoclonal | |
FITC | |
myeloid specific antigen, myeloid specific protein, p183 DNA-binding cytoplasmic ptotein | |
Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
BM-2 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title