Learn More
Myeloid Cell Marker, Mouse anti-Human,Primate, Alexa Fluor 532, Clone: BM-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP233114AF532
Catalog No. NB005649
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Myeloid Cell Marker Monoclonal antibody specifically detects Myeloid Cell Marker in Human, Primate samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Specifications
Myeloid Cell Marker
BM-2
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified, Conjugated
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Primate
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50mM Sodium Borate with 0.05% Sodium Azide
myeloid specific antigen, myeloid specific protein, p183 DNA-binding cytoplasmic ptotein
Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes were used as the immunogen for this granulocyte antibody.
Protein G purified
RUO
Primary
Recognizes an unidentified antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. MAb BM-2 reacts with early precursor and mature forms of human myeloid cells. This MAb is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes.