Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloid leukemia factor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | Myeloid leukemia factor 1 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB022507
|
Novus Biologicals
NBP293971100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB022508
|
Novus Biologicals
NBP29397120UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Myeloid leukemia factor 1 Polyclonal antibody specifically detects Myeloid leukemia factor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Myeloid leukemia factor 1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Myelodysplasia-myeloid leukemia factor 1, myeloid leukemia factor 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-268 of human MLF1 (NP_071888.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Stem Cell Markers | |
PBS (pH 7.3), 50% glycerol | |
4291 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title