Learn More
Cell Biolabs IncSupplier Diversity Partner MYELOPEROXIDASE ACT ASSY COLOR  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA803
OxiSelect Myeloperoxidase Chlorination Activity Assay Kit is a quantitative, colorimetric assay for measuring the chlorination activity of myeloperoxidase within a sample. 200 assays/kit including standards and unknowns.
Catalog No. 50-673-724