Learn More
BioVision IncSupplier Diversity Partner Myeloperoxidase (MPO) Colorimetric Activity Assay Kit: High-Throughput Suitable Assay for Measuring MPO Activity in various Samples such as Serum, Tissues, Cells etc. Detection Limit: 0.05 mU. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K744100
Myeloperoxidase (MPO) is a peroxidase enzyme (EC 1.11.1.7) most abundantly expressed in neutrophil granulocytes. It is a lysosomal protein stored in the azurophilic granules of the neutrophil. MPO contains a heme pigment which causes its green color in secretions rich in neutrophils, such as pus and some forms of mucus. MPO catalyzes the production of hypochlorous acid (HClO) from hydrogen peroxide (H₂O₂) and chloride anion (Cl-, or the equivalent from a non-chlorine halide). MPO also oxidizes tyrosine to a tyrosyl radical using hydrogen peroxide as an oxidizing agent. In BioVision’s MPO Assay Kit, the HClO produced from H₂O₂ and Cl- is reacted with taurine to generate the taurine chloramine, which subsequently reacts with the TNB2- probe to eliminate color (λ = 412 nm). The kit provides a rapid, simple, sensitive, and reliable test suitable for high throughput activity assay of MPO. This kit can be used to detect MPO as low as 0.05 mU per well.
Catalog No. 50-111-9872
Due to product restrictions, please to purchase or view availability for this product.