Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloperoxidase (MPO) Monoclonal Antibody (OTI11C7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF813994


Description
Myeloperoxidase (MPO) Monoclonal antibody specifically detects Myeloperoxidase (MPO) in Human samples. It is validated for ELISASpecifications
Myeloperoxidase (MPO) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P05164 | |
Mouse | |
Protein A/G | |
RUO | |
4353 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
ELISA | |
OTI11C7 | |
Unconjugated | |
MPO | |
84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
Full length recombinant protein of human MPO (NP_000241) produced in Baculovirus-Insect cell. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction