Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloperoxidase/MPO Mouse anti-Human, Alexa Fluor™ 532, Clone: MPO421-8B2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP241406AF532
Description
Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Flow Cytometry, ELISA, Flow (Intracellular).Specifications
Myeloperoxidase/MPO | |
MPO421-8B2 | |
Flow Cytometry, ELISA, Flow (Intracellular) | |
Purified, Conjugated | |
MPO | |
Human myeloperoxidase | |
Protein A purified | |
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
Primary | |
4353.0 | |
Human |
Flow Cytometry, ELISA | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
The mouse monoclonal antibody MPO421-8B2 recognizes human myeloperoxidase, a heme protein present in myeloblasts, neutrophils and monocytes. It is a marker of acute myelogenous leukemias and acute lymphoblastic leukemias. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title