Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase/MPO Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142591SS

 View more versions of this product

Catalog No. NBP142591SS

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Myeloperoxidase/MPO Polyclonal antibody specifically detects Antigen in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications

Specifications

Myeloperoxidase/MPO
Unconjugated
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3. with 0.05mg Thimerosal
Affinity Purified
MPO
A synthetic peptide corresponding to a sequence mapping at the C-terminal of human MPO, identical to the related mouse and rat sequence.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Please note the larger size(s) come in lyophilized form and the sample size will be in liquid formate. Reconstitute with deionized water.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 2 ug/ml, Immunohistochemistry 10.5-1 ug/ml, Immunohistochemistry-Paraffin 10.5-1 ug/ml
P05164
EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Rabbit
IgG
0.025 mg
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Primary
4353
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit