Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550239

Catalog No. PA550239


Add to cart

Description

Description

The antibody detects endogenous levels of total MYL2 protein.

Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.
Specifications

Specifications

MYL2
Polyclonal
Unconjugated
MYL2
Liquid
MYL2
Full length fusion protein of Human MYL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P10916, P51667
MLC2, CMH10
Rabbit
IgG
100 μL
-20°C
Primary
17906, 4633
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit