Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL4 Mouse anti-Human, Alexa Fluor™ 594, Clone: 400000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP242646AF594
Description
MYL4 Monoclonal specifically detects MYL4 in Human samples. It is validated for Western Blot, ELISA.Specifications
MYL4 | |
400000000 | |
Western Blot, ELISA | |
Purified, Conjugated | |
MYL4 | |
Recombinant human MYL4 (1-197aa) purified from E. coli | |
Protein A purified | |
Vision | |
Primary | |
4635.0 |
Western Blot, ELISA | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
ALC1, AMLC, GT1, MLC1, Myosin light chain 1, embryonic muscle/atrial isoform, myosin light chain 4, Myosin light chain alkali GT-1 isoform, myosin, atrial/fetal muscle, light chain, myosin, light chain 4, alkali; atrial, embryonic, myosin, light polypeptide 4, alkali; atrial, embryonic, PRO1957 | |
Mouse | |
IgG2a Kappa | |
100 μL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title