Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549205

Catalog No. PA549205


Add to cart

Description

Description

Regulatory light chain of myosin. Does not bind calcium.
Specifications

Specifications

MYL4
Polyclonal
PBS with 0.09% sodium azide
P12829
Myosin light chain 4, Myosin light chain 1, embryonic muscle/atrial isoform, Myosin light chain alkali GT-1 isoform, MYL4, MLC1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4635
Western Blot
Unconjugated
MYL4
Liquid
MYL4
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human MYL4.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit