Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL7, Mouse anti-Human, Clone: 4E7, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB16341

Catalog No. 89-999-099


Add to cart

Description

Description

Specifications

Specifications

MYL7
4E7
Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
myosin, light chain 7, regulatory
MYL2A/MYLC2A
Mouse
IgG1
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
Liquid
MYL7
Recombinant protein corresponding to full length human MYL7 from E. coli.
50 μL
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
58498
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit