Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL7 Monoclonal Antibody (OTI7F2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526389
Description
MYL7 Monoclonal antibody specifically detects MYL7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MYL7 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q01449 | |
Myl7 | |
Full length human recombinant protein of MYL7 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI7F2 | |
Unconjugated | |
Myl7 | |
MLC2a; MLC-2a; MLC-2alpha; MYL2A; MYL7; Mylc2a; myosin light chain 2a; myosin light chain 7; myosin light chain, regulatory A; myosin regulatory light chain 2, atrial isoform; Myosin regulatory light chain 7; myosin, light chain 7, regulatory; myosin, light polypeptide 7, regulatory; RLC-A | |
Mouse | |
Affinity Chromatography | |
RUO | |
58498 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction