Learn More
MYL9 Recombinant Protein, Abnova
Manufacturer:  Abnova Corporation H00010398P0110
 View more versions of this product
Catalog No. 89-011-870
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Human MYL9 Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)
Human
Solution
38.72
8
Glutathione Sepharose 4 Fast Flow
10μg
-80°C
Yes