Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYLIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542302
Description
Peptide sequence: CSSCEGLSCQ QTRVLQEKLR KLKEAMLCMV CCEEEINSTF CPCGHTVCCE Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
The ERM protein family members ezrin, radixin, and moesin are cytoskeletal effector proteins linking actin to membrane-bound proteins at the cell surface. Myosin regulatory light chain interacting protein (MYLIP) is a novel ERM-like protein that interacts with myosin regulatory light chain and inhibits neurite outgrowth.
Specifications
MYLIP | |
Polyclonal | |
Unconjugated | |
MYLIP | |
9430057C20Rik; band 4.1 superfamily member BZF1; BM-023; BZF1; cellular modulator of immune recognition (c-MIR); E3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor; E3 ubiquitin-protein ligase MYLIP; I79_001311; IDOL; inducible degrader of the LDL-receptor; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase MYLIP; MIR; Mylip; Myosin regulatory light chain interacting protein; myosin regulatory light chain-interacting protein; PP5242; RING-type E3 ubiquitin transferase MYLIP | |
Rabbit | |
Affinity chromatography | |
RUO | |
29116 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WY64 | |
MYLIP | |
Synthetic peptide directed towards the middle region of human MYLIP. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction