Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYLIP Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154551AP

Catalog No. 50-173-0241


Add to cart

Description

Description

The ERM protein family members ezrin, radixin, and moesin are cytoskeletal effector proteins linking actin to membrane-bound proteins at the cell surface. Myosin regulatory light chain interacting protein (MYLIP) is a novel ERM-like protein that interacts with myosin regulatory light chain and inhibits neurite outgrowth.
Specifications

Specifications

MYLIP
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZDI6, Q8BM54, Q8WY64
BZF1, Idol, MIR, MYLIP, PP5242
Rabbit
IgG
150 μL
-20°C
Primary
100760940, 218203, 29116, 306825
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MYLIP
Liquid
MYLIP
MYLIP Fusion Protein Ag7720
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit