Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYLK2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550236

Catalog No. PA550236


Add to cart

Description

Description

The antibody detects endogenous levels of total MYLK2 protein.

This gene encodes a myosin light chain kinase, a calcium/calmodulin dependent enzyme, that is exclusively expressed in adult skeletal muscle.
Specifications

Specifications

MYLK2
Polyclonal
Unconjugated
MYLK2
Liquid
MYLK2
Fusion protein corresponding to residues near the C terminal of human myosin light chain kinase 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9H1R3, Q8VCR8
KMLC, MLCK, MLCK2, skMLCK
Rabbit
IgG
100 μL
-20°C
Primary
228785, 85366
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit