Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYNN Monoclonal Antibody (OTI1F10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526393
Description
MYNN Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes a member of the BTB/POZ and zinc finger domain-containing protein family that are involved in the control of gene expression. Alternative splicing results in multiple transcript variants and a pseudogene has been identified on chromosome 14.
Specifications
MYNN | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NPC7 | |
MYNN | |
Human recombinant protein fragment corresponding to amino acids 10-300 of MYNN produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1F10 | |
Unconjugated | |
MYNN | |
2810011C24Rik; AA415053; AI661031; AW049392; MYNN; Mynr; myoneurin; OSZF; SBBIZ1; ZBTB31; Zinc finger and BTB domain-containing protein 31; zinc finger protein with BTB/POZ domain; ZNF902 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55892 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction