Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYO1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 150121AP150UL
Description
Motor protein that may participate in process critical to neuronal development and function such as cell migration, neurite outgrowth and vesicular transport.Specifications
MYO1B | |
Polyclonal | |
Unconjugated | |
MYO1B | |
MMI alpha, MMIa, MYH 1c, MYO1B, Myosin I alpha, myosin IB, myr1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117057, 17912, 4430 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43795, P46735, Q05096 | |
Myo1b | |
MYO1B Fusion Protein Ag6895 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction