Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYO1C Monoclonal Antibody (221CT4.7.2.4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537513
Description
MYO1C Monoclonal Antibody for Western Blot
MYO1C is a member of the unconventional myosin protein family, which are actin-based molecular motors. The protein is found in the cytoplasm, and one isoform with a unique N-terminus is also found in the nucleus. The nuclear isoform associates with RNA polymerase I and II and functions in transcription initiation. The mouse ortholog of this protein also functions in intracellular vesicle transport to the plasma membrane.
Specifications
MYO1C | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.09% sodium azide; pH 7.4 | |
O00159 | |
MYO1C | |
MYO1C recombinant protein. | |
RUO | |
4641 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
221CT4.7.2.4 | |
Unconjugated | |
MYO1C | |
C80397; FLJ23903; mm1beta; MMIb; MMI-beta; myo1c; myo1ca; MYO1E; myosin 1c, paralog a; myosin heavy chain myr 2; myosin I; Myosin I beta; myosin IB; myosin IC; myosin Ic, paralog a; myosin IC-like; myosin-I beta; myosin-Ibeta; myosin-Ic; Myr2; NMI; nuclear myosin I; nuclear myosin I beta; si:ch211-69i14.4; unconventional myosin Myr2 I heavy chain; unconventional myosin-Ic | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction