Learn More
MYO1E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 177681AP
Catalog No. 50-173-1779
Add to cart
Description
Description
Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments.
Specifications
Specifications
MYO1E
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E9Q634, Q12965, Q63356
HuncM IC, MYO1C, MYO1E, Myosin Ic, myosin IE
Rabbit
IgG
150 μL
-20°C
Primary
25484, 4643, 71602
Western Blot
Unconjugated
MYO1E
Liquid
MYO1E
MYO1E Fusion Protein Ag12176
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat