Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MyoD Mouse anti-Human, Clone: rMYD712, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30739420UG
Description
MyoD Monoclonal specifically detects MyoD in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
MyoD | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
Recombinant full-length human MyoD protein | |
20 μg | |
Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific | |
4654 | |
Store at 4°C. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Peptide Array | |
rMYD712 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction