Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyoD Rabbit anti-Human, Clone: MYOD1/3418R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen MyoD
Clone MYOD1/3418R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122282
View Documents
Novus Biologicals
NBP30740420UG
20 μg
Each for $159.82
Add to cart
 
NB122281
View Documents
Novus Biologicals
NBP30740410UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

MyoD Monoclonal antibody specifically detects MyoD in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MyoD
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM
Synthetic peptide corresponding to residues within (aa1-100) of human MyoD
Monoclonal
Protein A purified
MYOD1/3418R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific
10 mM PBS with 0.05% BSA
4654
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit