Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyoD, Rabbit anti-Human, DyLight 350, Clone: MYOD1/2075R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275772UV

Catalog No. NB009092


Add to cart

Description

Description

MyoD Monoclonal antibody specifically detects MyoD in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

MyoD
MYOD1/2075R
Immunohistochemistry, Immunohistochemistry-Paraffin
50mM Sodium Borate with 0.05% Sodium Azide
MYOD1
Recombinant full-length human MyoD protein
Monoclonal
Protein A purified
RUO
Store at 4°C in the dark.
Recognizes a phospho-protein of 45kDa, identified as MyoD. This MAb does not cross react with myogenin, Myf5, or Myf6. Antibody to MyoD labels the nuclei of myoblasts in developing muscle tissues. MyoD is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occasionally nuclear expression of MyoD is seen in ectomesenchymoma and a subset of Wilm s tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing s sarcomas and alveolar soft part sarcomas. Only nuclear staining should be considered as evidence of skeletal muscle differentiation.
Immunohistochemistry,Immunohistochemistry (Paraffin)
DyLight 350
Purified,Conjugated
bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM
Rabbit
IgG
Primary
0.1 mL
Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit