Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MyoD, Rabbit anti-Human, Janelia Fluor 549, Clone: MYOD1/2075R, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275772JF549
Description
MyoD Monoclonal antibody specifically detects MyoD in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
MyoD | |
MYOD1/2075R | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MYOD1 | |
Recombinant full-length human MyoD protein | |
Monoclonal | |
Protein A purified | |
RUO | |
Store at 4°C in the dark. | |
Recognizes a phospho-protein of 45kDa, identified as MyoD. This MAb does not cross react with myogenin, Myf5, or Myf6. Antibody to MyoD labels the nuclei of myoblasts in developing muscle tissues. MyoD is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occasionally nuclear expression of MyoD is seen in ectomesenchymoma and a subset of Wilm s tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing s sarcomas and alveolar soft part sarcomas. Only nuclear staining should be considered as evidence of skeletal muscle differentiation. |
Immunohistochemistry,Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
Purified,Conjugated | |
bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
Rabbit | |
IgG | |
Primary | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title