Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myofibroblast Marker Mouse anti-Human, Clone: PR 2D3, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Myofibroblast Marker |
---|---|
Clone | PR 2D3 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122964
|
Novus Biologicals
NBP30774520UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Myofibroblast Marker Monoclonal specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
Myofibroblast Marker | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Crude homogenate of normal human colorectal mucosa | |
Primary | |
Store at 4°C. |
PR 2D3 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title