Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Myogenin Ab-1 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Deliver accurate, reproducible results with the Epredia™ Myogenin Ab-1, Mouse Monoclonal Antibody.

$734.00 - $1141.00

Specifications

Antigen Myogenin Ab-1
Clone F5D
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1113P1ABX
View Documents
Epredia™
MS1113P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $734.00
Add to cart
 
MS1113PABX
View Documents
Epredia™
MS1113PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $1,141.00
Add to cart
 
MS1113R1
View Documents
Epredia™
MS1113R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
MS1113R7
View Documents
Epredia™
MS1113R7
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Myogenin is a member of a family of myogenic regulatory genes, which includes MyoD, myf5 and MRF4. These genes encode a set of transcription factors, which are essential for muscle development. Transfection of myogenin into multipotential mesodermal cells have been shown to convert the mesodermal cells to myoblasts. Expression of myogenin is restricted to cells of skeletal muscle origin. It is therefore a useful marker for tumors of the muscle lineage, being strongly expressed in alveolar rhabdomyosarcomas.

Host Species: Mouse

Clone: F5D

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat and Cat. Does not react with quail. Others not known.

Epitope: aa 138-158

Immunogen: Recombinant protein containing rat myogenin aa 30-224

Molecular Weight: 34kDa

Positive Control: Rh30 cells, Rhabdomyosarcoma or Wilm's tumors

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Myogenin Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Primary
IgG2a kappa
F5D
Monoclonal
Mouse
Muscle Biology
Recombinant protein containing rat myogenin aa 30-224
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit