Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Myoglobin, Rabbit Polyclonal Antibody

Provides accurate, reproducible results with the Thermo Scientific™ Myoglobin, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB9068P

 View more versions of this product

Catalog No. RB9068P

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provides accurate, reproducible results with the Thermo Scientific™ Myoglobin, Rabbit Polyclonal Antibody.

Myoglobin is a cytoplasmic, single chain polypeptide of 153 amino acids and contains a single heme group. Myoglobin is found in skeletal and carciac muscle but not in smooth muscle and functions as an oxygen transporting pigment. Upon damage to the muscle cell due to infarction of a coronary artery, neurological trauma, infection of tumor processes, myoglobin escapes to the environment and can be found in plasma using sensitive assays.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Purified human myoglobin

Molecular Weight: 17.8kDa

Positive Control: Skeletal muscle

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Myoglobin
Polyclonal
200µg/mL
Concentrated
Purified human myoglobin
1mL
Muscle Biology
Primary
Human
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
Rabbit
IgG
RUO
Primary
Polyclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit