Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myomegalin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530324
Description
Recommended positive controls: NT2D1, PC-3, SK-N-SH, mouse brain. Predicted reactivity: Human (98%), Mouse (83%), Rat (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene serves to anchor phosphodiesterase 4D to the Golgi/centrosome region of the cell. Defects in this gene may be a cause of myeloproliferative disorder (MBD) associated with eosinophilia. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Myomegalin | |
Polyclonal | |
Unconjugated | |
PDE4DIP | |
4732458A06Rik; 9430063L05Rik; C87016; cardiomyopathy-associated protein 2; CMYA2; D130016K21Rik; D3Bwg1078e; KIAA0454; KIAA0477; mKIAA0454; MMGL; Myomegalin; myomegalin/phosphodiesterase 4D interacting protein variant 8; Pbp46; PDE4DIP; phosphodiesterase 4D interacting protein; phosphodiesterase 4D interacting protein (myomegalin); phosphodiesterase 4D-interacting protein; Phosphodiesterase-binding protein clone 46; upregulated during skeletal muscle growth 4; Usmg4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83679, 9659 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.81 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q5VU43, Q80YT7 | |
PDE4DIP | |
Recombinant protein fragment corresponding to a region within amino acids 99 and 283 of Human Myomegalin. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction