Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin heavy chain 11 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259221

Catalog No. NBP259221


Add to cart

Description

Description

Myosin heavy chain 11 Polyclonal antibody specifically detects Myosin heavy chain 11 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA.
Specifications

Specifications

Myosin heavy chain 11
Unconjugated
Serum with 0.01% Sodium Azide
AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain
Rabbit
IgG
100 ul
Hypoxia, Stem Cell Markers
Primary
4629
Western Blot, ChIP assay, ELISA
Western Blot 1:1000, Chromatin Immunoprecipitation 5 ul, ELISA 1:200
Antiserum
MYH11
The exact immunogen is propietary information.
Whole Antiserum
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only