Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Light Chain 2 Mouse anti-Human, Mouse, Xenopus, PE, Clone: 3B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130249PE

Catalog No. NBP130249PE


Add to cart

Description

Description

Myosin Light Chain 2 Monoclonal antibody specifically detects Myosin Light Chain 2 in Human, Mouse, Xenopus samples. It is validated for Flow Cytometry.
Specifications

Specifications

Myosin Light Chain 2
3B2
Flow Cytometry
Purified
MYL2
Recombinant human MYL2 (1-166aa) purified from E. coli (NP_000423).
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MYL2 protein.
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2
Mouse
IgG2b Kappa
0.1 ml
Phospho Specific
Primary
4633
Human, Mouse, Xenopus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit