Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin Light Chain 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$176.85 - $436.00
Specifications
Concentration | 1.0 mg/mL |
---|---|
Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10082061
|
Novus Biologicals
NB10082061 |
0.1 mL |
Each for $436.00
|
|
NB017862
|
Novus Biologicals
NB100820610.025ML |
0.025 mL |
Each for $176.85
|
|
Description
Myosin Light Chain 2 Polyclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
P10916, P08733 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
IgG | |
Affinity Purified | |
18 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title