Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin Phosphatase Rabbit anti-Human, Chicken, Clone: SA19, Novus Biologicals™
Rabbit Monoclonal Antibody
$176.84 - $666.00
Specifications
Antigen | Myosin Phosphatase (p Thr853) |
---|---|
Clone | SA19 |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Myosin Phosphatase Monoclonal specifically detects Myosin Phosphatase in Human, Chicken samples. It is validated for Western Blot.Specifications
Myosin Phosphatase (p Thr853) | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Rabbit | |
Phospho Specific, Protein Phosphatase | |
No buffer with No Preservative | |
MBSmyosin phosphatase, target subunit 1, MGC133042, Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, MYPT1protein phosphatase 1 regulatory subunit 12A, protein phosphatase 1, regulatory (inhibitor) subunit 12A, Protein phosphatase myosin-binding subunit | |
PPP1R12A | |
IgG | |
Affinity Purified | |
MYPT1 phosphorylated on Thr850 doublet at 130kDa weight may be detected. |
SA19 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Chicken | |
O14974 | |
4659 | |
KLH-conjugated, synthetic peptide containing the sequence.RS[pT]GV., in which [pT] corresponds to phosphorylated threonine at amino acid residue 850 of chicken myosin phosphatase target subunit 1 (MYPT1). Corresponds to Thr853 in human MYPT1. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title