Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody

Provides accurate, reproducible results with the Thermo Scientific™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS1348P1

 View more versions of this product

Catalog No. MS1348P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provides accurate, reproducible results with the Thermo Scientific™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody.

SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.

Host Species: Mouse

Clone: SM-M10

Isotype: IgG1/κ

Species Reactivity: Human and Cow. Others not known.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 205kDa (MHC-1) and 200kDa (MHC-2)

Positive Control: Uterus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence
  • Immunoprecipitation (Not verified)
Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Myosin, Smooth Muscle Heavy Chain Ab-3
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG2a kappa
RUO
Primary
Bovine, Human
Immunofluorescence, Immunohistochemistry (Paraffin)
SM-M10
Unconjugated
Concentrated
Crude human uterus extract
500μL
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit