Learn More
Myosinheavychain2 Mouse anti-Human, Janelia Fluor 646, Clone: TH81 (BGN/04/4481), Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10065675JF646
Catalog No. NB065675646
Add to cart
Description
Description
Myosinheavychain2 Monoclonal antibody specifically detects Myosinheavychain2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Myosinheavychain2
TH81 (BGN/04/4481)
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
MYH2
Myosin heavy chain from rabbit muscle
Protein A purified
Cytoskeleton Markers
Primary
4620
Human, Rabbit, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
fast 2a myosin heavy chain, MYH2A, MYHas8, MyHC-2A, MyHC-IIa, MYHSA2, myosin heavy chain 2, myosin heavy chain 2a, myosin heavy chain IIa, myosin, heavy chain 2, skeletal muscle, adult, myosin, heavy polypeptide 2, skeletal muscle, adult, myosin-2, skeletal muscle, adult 2, type IIA myosin heavy chain
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
NB100-65675 reacts strongly with human cardiac myosin and also recognizes rabbit myosin heavy and light chains.