Learn More
MYOT Mouse anti-Human, Clone: OTI10A11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809553
Catalog No. 50-166-9581
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
Specifications
MYOT
Monoclonal
Unconjugated
MYOT
Lyophilized
MYOT
Full length human recombit protein of human MYOT produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI10A11
PBS with 8% trehalose and no preservative; pH 7.3
Q9UBF9
LGMD1, LGMD1A, MFM3, TTID, TTOD
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9499