Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myotrophin Monoclonal Antibody (1F3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524520
Description
Peptide Sequence: MCDKEFMWAL KNGGLDEVKD YVAKGEDVNR TLEGGRKPLH YAADCGQLEI LEFLLLKGAD INAPDKHHIT PLLSAVYEGH VSCVKLLLSK GADKTVKGPD GLTAFEATDN QAIKALLQ
Myotrophin Monoclonal antibody specifically detects Myotrophin in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Myotrophin | |
Monoclonal | |
Unconjugated | |
MTPN | |
5033418D15Rik; GCDP; Granule cell differentiation protein; growth factor; Mtpn; Myotrophin; protein V-1; stimulates myocyte growth; V1; V-1; V-1 protein | |
Mouse | |
Protein A | |
RUO | |
136319 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1F3 | |
PBS with no preservative; pH 7.4 | |
P58546 | |
MTPN | |
MTPN (AAH28093, 1 a.a. ∽ 118 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction