Learn More
MYOZ3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 238801AP
Catalog No. 50-173-4149
Add to cart
Description
Description
The protein encoded by this gene is specifically expressed in the skeletal muscle, and belongs to the myozenin family. Members of this family function as calcineurin-interacting proteins that help tether calcineurin to the sarcomere of cardiac and skeletal muscle. They play an important role in modulation of calcineurin signaling.
Specifications
Specifications
MYOZ3
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R4E4, Q8TDC0
Calsarcin 3, CS 3, CS3, FATZ related protein 3, FRP3, MYOZ3, myozenin 3
Rabbit
IgG
150 μL
-20°C
Primary
170947, 689274, 91977
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MYOZ3
Liquid
Myoz3
MYOZ3 Fusion Protein Ag20839
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat