Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myricetin 95%, ACROS Organics™

Click to view available options
Quantity:
25mg
Packaging:
Glass bottle
529-44-2
C15H10O8
318.237
MFCD00006827
IKMDFBPHZNJCSN-UHFFFAOYSA-N
myricetin,cannabiscetin,myricetol,myricitin,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one,3,3',4',5,5',7-hexahydroxyflavone,3,5,7,3',4',5'-hexahydroxyflavone,unii-76xc01ftoj,ccris 5838,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
5281672
CHEBI:18152
3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
C1=C(C=C(C(=C1O)O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O
529-44-2
C15H10O8
318.237
MFCD00006827
IKMDFBPHZNJCSN-UHFFFAOYSA-N
myricetin,cannabiscetin,myricetol,myricitin,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one,3,3',4',5,5',7-hexahydroxyflavone,3,5,7,3',4',5'-hexahydroxyflavone,unii-76xc01ftoj,ccris 5838,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
5281672
CHEBI:18152
3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
C1=C(C=C(C(=C1O)O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown-Green to Brown-Yellow
Quantity 25mg
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Beilstein 18, 257
Merck Index 15, 6417
Formula Weight 318.23
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material Myricetin, 95%
Show More Show Less