Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myristoyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
112-64-1
C14H27ClO
246.82
MFCD00000741
LPWCRLGKYWVLHQ-UHFFFAOYSA-N
myristoyl chloride,myristoylchloride,tetradecanoylchloride,e r eao poundae,myristic acid chloride,tetradecanoic acid chloride,acmc-2099fr,ksc181q3r
66986
tetradecanoyl chloride
CCCCCCCCCCCCCC(=O)Cl
112-64-1
C14H27ClO
246.82
MFCD00000741
LPWCRLGKYWVLHQ-UHFFFAOYSA-N
myristoyl chloride,myristoylchloride,tetradecanoylchloride,e r eao poundae,myristic acid chloride,tetradecanoic acid chloride,acmc-2099fr,ksc181q3r
66986
tetradecanoyl chloride
CCCCCCCCCCCCCC(=O)Cl
Boiling Point 172°C to 176°C (5.0 mbar)
Melting Point -1°C
Flash Point >100°C
Refractive Index 1.4480 to 1.45
Quantity 5g
Infrared Spectrum Authentic
Color Undesignated
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)12COCl
Beilstein 02, 368
Solubility Information Solubility in water: may decompose
Formula Weight 246.82
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Myristoyl chloride
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May form explosive peroxides.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 203-994-6