Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Myristoylcarnitine chloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
14919-38-1
C21H42ClNO4
408.02
TWGWHMYOGIGWDM-UHFFFAOYSA-N
+/--myristoylcarnitine chloride,a+/--myristoylcarnitine chloride,3-carboxy-2-tetradecanoyloxy propyl trimethylazanium chloride,3-carboxy-n,n,n-trimethyl-2-tetradecanoyloxy propan-1-aminium chloride
24802347
(3-carboxy-2-tetradecanoyloxypropyl)-trimethylazanium;chloride
CCCCCCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]
This item is not returnable. View return policy
14919-38-1
C21H42ClNO4
408.02
TWGWHMYOGIGWDM-UHFFFAOYSA-N
+/--myristoylcarnitine chloride,a+/--myristoylcarnitine chloride,3-carboxy-2-tetradecanoyloxy propyl trimethylazanium chloride,3-carboxy-n,n,n-trimethyl-2-tetradecanoyloxy propan-1-aminium chloride
24802347
(3-carboxy-2-tetradecanoyloxypropyl)-trimethylazanium;chloride
CCCCCCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]

Intermediate in lipid metabolism

Homolog of acetylcarnitine chloride (Cat. No. 0355). Acylcarnitines are important intermediates in lipid metabolism.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 408.02
Percent Purity >97%
Chemical Name or Material (±)-Myristoylcarnitine chloride

Recommended Storage : Store at -20°C